Learn More
Uvex Micro Black/Orange PU/RU 65284 Low Shoe - Width 14
Incorporates shock absorption, ergonomic design and low weight materials to ensure max performance, lightweight PU midsole with VIBRAM rubber outsole for high levels of cut, chemical and heat resistance, improved grip on slippery terrain
1770.00 SEK
Specifications
Color | Black, Orange |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane rubber (PU/RU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17241431
|
Uvex
6528435 |
35 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17251431
|
Uvex
6528436 |
36 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17261431
|
Uvex
6528437 |
37 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17271431
|
Uvex
6528438 |
38 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17281431
|
Uvex
6528439 |
39 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17291431
|
Uvex
6528440 |
40 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17201441
|
Uvex
6528441 |
41 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17211441
|
Uvex
6528442 |
42 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17221441
|
Uvex
6528443 |
43 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17231441
|
Uvex
6528444 |
44 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17241441
|
Uvex
6528445 |
45 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17251441
|
Uvex
6528446 |
46 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17261441
|
Uvex
6528447 |
47 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17271441
|
Uvex
6528448 |
48 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17281441
|
Uvex
6528449 |
49 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17291441
|
Uvex
6528450 |
50 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17201451
|
Uvex
6528451 |
51 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
17211451
|
Uvex
6528452 |
52 |
1770.00 SEK
1 pair |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 VIBRAM Shoe 6528.4 S3 HI HRO SRC
- Sporty, multifunctional safety shoes
- Extremely light and flexible
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side and ankle twist stability, no thermal conductivity and anti-magnetic properties
- Metal-free, penetration-resistance insole in accordance with latest standard, which does not impact shoe flexibility
- Made with new high-performance technology for exceptional wearer comfort
- Uvex Climazone technology for optimum comfort
- Breathable, water-repellent high-tech microvelour upper material
- Distance mesh lining provides optimum breathability
- Made from synthetic textiles, suitable for those allergic to chrome
- Reflecting elements for more safety
- Individually adjustable due to elastic laces with quick-locking mechanism
- Normal laces also provided
- Virtually seam-free construction to eliminate pressure points
- Removable comfort insole, antistatic, with moisture control system and additional shock absorption in heel and at front of foot
- Soft, padded collar and tongue, dust tongue
- Injection-molded polyurethane scuffcap to protect upper leather on toe cap and significantly extend resistance time
- Meets ESD requirements: resistance to ground Kleiner
- Lightweight two-layer insole with excellent shock absorption characteristics and outstanding slip resistance
- Multilayer sole system comprising PU midsole and highly slip resistant rubber outersole from VIBRAM
- Optimized ankle protection to ensure safety when working on ladders
- Rugged, self-cleaning sole ridges ensure max grip for demanding applications on uneven or dirty floors (SRC labeling)
- Withstands brief exposure to heat approx. 300°C
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6528.4
- Standard EN ISO 20345:2011 S3 HI HRO SRC
- Sizes: 35 to 52
Specifications
Black, Orange | |
Dual-density polyurethane rubber (PU/RU) | |
No | |
Unisex | |
Chemicals and heat | |
Yes | |
S3 |
Microvelour | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Uvex 2 | |
No | |
SRC |