Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
Brand:  ROBU VitraPOR™ 18032
Additional Details : Weight : 0.07410kg
 View more versions of this product
Product Code. 16802926
  • 2990.00 SEK / Pack of 5
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
2
20 mm
Non-sterile
30 mm