Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
Brand: ROBU VitraPOR™ 18033
Additional Details : Weight : 0.07410kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
3 | |
20 mm |
Non-sterile | |
30 mm |