Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
Brand:  ROBU VitraPOR™ 18004
Additional Details : Weight : 0.02140kg
 View more versions of this product
Product Code. 16842906
  • 1960.00 SEK / Pack of 5
Estimated Shipment: 06-05-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
4
6 mm
Non-sterile
15 mm