Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
Brand:  ROBU VitraPOR™ 18010
Additional Details : Weight : 0.02520kg
 View more versions of this product
Product Code. 16862906
  • 1785.00 SEK / Pack of 5
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
0
9 mm
Non-sterile
20 mm