Learn More
ROBU VitraPOR™ Micro-Filter Candle, Cylindrical Shape
Micro-Filter Candle without glass tube.
Brand: ROBU VitraPOR™ 18044
Additional Details : Weight : 0.10740kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
4 | |
20 mm |
Non-sterile | |
55 mm |