Learn More
ROBU VitraPOR™ Micro-Filter Candle with Glass Tube
Micro-Filter Candle with narrow glass tube.
2285.00 SEK - 5870.00 SEK
Specifications
Material Borosilicate Glass (3.3)
Sterility Non-sterile
Includes Glass Tube
View More Specs
Products 19
Product Code Brand Diameter (Metric) Length (Metric) Porosity Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Porosity Price Quantity & Availability  
16842936
View Documents Promotion Details
ROBU VitraPOR™
18111
9 mm 20 mm 1
2285.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16812946
View Documents Promotion Details
ROBU VitraPOR™
18122
13 mm 25 mm 2
2285.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16842946
View Documents Promotion Details
ROBU VitraPOR™
18125
13 mm 25 mm 5
5870.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16832936
View Documents Promotion Details
ROBU VitraPOR™
18110
9 mm 20 mm 0
2285.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16832956
View Documents Promotion Details
ROBU VitraPOR™
18143
20 mm 55 mm 3
4610.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16832946
View Documents Promotion Details
ROBU VitraPOR™
18124
13 mm 25 mm 4
2515.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16862946
View Documents Promotion Details
ROBU VitraPOR™
18131
20 mm 30 mm 1
3870.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16892936
View Documents Promotion Details
ROBU VitraPOR™
18120
13 mm 25 mm 0
2285.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16882936
View Documents Promotion Details
ROBU VitraPOR™
18115
9 mm 20 mm 5
4850.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16822936
View Documents Promotion Details
ROBU VitraPOR™
18105
6 mm 15 mm 5
3955.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16842956
View Documents Promotion Details
ROBU VitraPOR™
18144
20 mm 55 mm 4
5255.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16812956
View Documents Promotion Details
ROBU VitraPOR™
18141
20 mm 55 mm 1
4610.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16822956
View Documents Promotion Details
ROBU VitraPOR™
18142
20 mm 55 mm 2
4610.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16892946
View Documents Promotion Details
ROBU VitraPOR™
18134
20 mm 30 mm 4
4320.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16802936
View Documents Promotion Details
ROBU VitraPOR™
18102
6 mm 15 mm 2
2285.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16872946
View Documents Promotion Details
ROBU VitraPOR™
18132
20 mm 30 mm 2
3870.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16802956
View Documents Promotion Details
ROBU VitraPOR™
18140
20 mm 55 mm 0
4610.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16852946
View Documents Promotion Details
ROBU VitraPOR™
18130
20 mm 30 mm 0
3870.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
16882926
View Documents Promotion Details
ROBU VitraPOR™
18100
6 mm 15 mm 0
2285.00 SEK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
Non-sterile
Glass Tube