Learn More
ROBU VitraPOR™ Micro-Filter Candle with Glass Tube
Micro-Filter Candle with narrow glass tube.
2285.00 SEK - 5870.00 SEK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Sterility | Non-sterile |
Includes | Glass Tube |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Porosity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Porosity | Price | Quantity & Availability | |||
16842936
|
ROBU VitraPOR™
18111 |
9 mm | 20 mm | 1 |
2285.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16812946
|
ROBU VitraPOR™
18122 |
13 mm | 25 mm | 2 |
2285.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16842946
|
ROBU VitraPOR™
18125 |
13 mm | 25 mm | 5 |
5870.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16832936
|
ROBU VitraPOR™
18110 |
9 mm | 20 mm | 0 |
2285.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16832956
|
ROBU VitraPOR™
18143 |
20 mm | 55 mm | 3 |
4610.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16832946
|
ROBU VitraPOR™
18124 |
13 mm | 25 mm | 4 |
2515.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16862946
|
ROBU VitraPOR™
18131 |
20 mm | 30 mm | 1 |
3870.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16892936
|
ROBU VitraPOR™
18120 |
13 mm | 25 mm | 0 |
2285.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16882936
|
ROBU VitraPOR™
18115 |
9 mm | 20 mm | 5 |
4850.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16822936
|
ROBU VitraPOR™
18105 |
6 mm | 15 mm | 5 |
3955.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16842956
|
ROBU VitraPOR™
18144 |
20 mm | 55 mm | 4 |
5255.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16812956
|
ROBU VitraPOR™
18141 |
20 mm | 55 mm | 1 |
4610.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16822956
|
ROBU VitraPOR™
18142 |
20 mm | 55 mm | 2 |
4610.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16892946
|
ROBU VitraPOR™
18134 |
20 mm | 30 mm | 4 |
4320.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16802936
|
ROBU VitraPOR™
18102 |
6 mm | 15 mm | 2 |
2285.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16872946
|
ROBU VitraPOR™
18132 |
20 mm | 30 mm | 2 |
3870.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16802956
|
ROBU VitraPOR™
18140 |
20 mm | 55 mm | 0 |
4610.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16852946
|
ROBU VitraPOR™
18130 |
20 mm | 30 mm | 0 |
3870.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
16882926
|
ROBU VitraPOR™
18100 |
6 mm | 15 mm | 0 |
2285.00 SEK
Pack of 5 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Non-sterile | |
Glass Tube |