Learn More
Eppendorf™ Micro Fusion Chamber
Easy to use. Integrated cuvette chamber and extremely high transformation rates
Brand:  Eppendorf™ 4308031000
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 15204566
  • 3878.00 SEK / Each
Estimated Shipment
07-06-2021
Description
Description
  • Extremely high transformation rates due to innovative electronics and buffers
  • Integrated cuvette chamber
  • Easy to use
  • Compact design
  • Universal electroporator for eukaryotic, yeast and bacteria
Specifications
Specifications
1Each
1
0.5mm
Micro Fusion Chamber