Läs mer
Thermo Scientific™ Micro Hot Plate, Ambient to 371°C, Aluminum
Produktkod. p-9034039 Handla allt Thermo Scientific Produkter
Spänning:
240 V
Förpackningsstorlek:
Varje
Produktkod. 15383757
Brand: Thermo Scientific™ HP2310BQ
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Shape: Round; Size: 3.5 in. dia.
Process delicate materials at low temperatures. The Thermo Scientific™ Micro Hotplate offers a compact, benchtop design with variable control, for quick sample testing.
  • Infinitely adjustable, demand-type control provides ±5°C (±9°F) temperature control over the entire operating range
  • Reaches 260°C (500°F) in just 4.5 minutes; 371°C (700°F) in 7.5 minutes
  • Cast-aluminum top plate
  • Embedded heating elements
  • Perforated stainless-steel case
  • Height: 3.75 in. (9.5 cm)
  • Top plate diameter: 3.5 in. (9 cm)
  • Weight: 4 lb. (1.8 kg)
TRUSTED_SUSTAINABILITY
Specifikationer
Material för toppplatta Aluminum
Storlek 3.5 in. dia.
Temperatur (metrisk) Ambient to 371°C
Höjd (engelska) 3.75 in.
Höjd (metrisk) 9.5 cm
Wattal 325 w
Elektriska krav 240 V, 50/60 Hz
Frekvens 50/60 Hz
Max. Provladdning 1.8 kg
Form Round
Spänning 240 V
Typ Hot Plate
Certifikat och efterlevnad UL Listed