Learn More
3M™ Microbial Luminescence System Injector Cleaning Kit GH620522587
Used for routine cleaning of the 3M™ Microbial Luminescence System II (MLS II)
Brand:  3M™ 7000043165
Additional Details : Weight : 0.34500kg
Product Code. 17268530
  • 2300.20 SEK / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
  • Used with the 3M™ Microbial Luminescence System II (MLS II)
  • Removes protein residues and microbial contamination in the injectors and reagent lines, thus avoiding high background readings, false positive results, and inaccurate dispensation of reagents.
Specifications
Specifications
3 M™ Microbial Luminescence System II (MLS II)