Learn More
3M™ Microbial Luminescence System Microwell Strip 3008
Microwell strip for use with the 3M™ Microbial Luminescence System (MLS).
Brand: 3M™ 7000030320
Additional Details : Weight : 0.48500kg
Description
- Plastic strips of 12 wells.
- Snap to desired number of wells.
- Use with 3M™ MLS Microwell Strip Holder #3009.
- Used primarily for reagent checks and low-volume UHT testing.
Specifications
Plastic Strips of 12 Wells | |
320/Cs. | |
3M™ MLS Microwell Strip Holder #3009. |