Learn More
Merck Millipore Microfilm Funnel and Filter
Convenient, easy to use system
Brand:  Merck Millipore MIHCWG250
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11973866
  • 3481.00 SEK / Pack of 150
Estimated Shipment: 20-08-2021
Add to basket
Description
Description
  • Convenient, easy to use system for well accepted membrane filtration method conforms to international standards: CEN, Standard Methods, WHO
  • Disposable, stackable, light, recyclable plastic Microfil™ funnels eliminate autoclaving and minimise waste
  • 'Push fit' Microfil™ funnels tightly seal to manifold supports without clamps or O-rings assuring leak-free operation and uniform micro-organism recoveries
  • Novel device lifts membrane after filtration, for easy removal with forceps
  • Unique Microfil™ support filters faster than all-glass filter apparatus
  • 100mL or 250mL funnels available with convenient accessory dispensers
  • Choose conventional sterile membranes or novel EZ-Pak™ membrane cartridges. Select manifold-mounted filtration supports for laboratory use or stand-alone on-site filtration support.
Specifications
Specifications
White Gridded
150 Pack
250 mL
150Pack
Polypropylene, Mixed Cellulose Ester