Learn More
Cytiva (formerly Pall Lab) MicroFunnel™ Filters Funnel with GN-6 Membrane
Ready-to-use, sterile and individually packaged MicroFunnel filter funnels reduce chances of cross contamination.
Brand: Cytiva (formerly Pall Lab) 4815
2421.79 SEK valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Additional Details : Weight : 1.39200kg
Description
- Easy squeeze removal of funnel to access membrane – no twisting means less RSI and membrane tearing
- Proven membrane performance
- For research use only
Specifications
300 mL | |
20/Pk. | |
Individually Wrapped |
0.45 μm | |
Sterile | |
GN-6 Metricel Mixed Cellulose Ester |