missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Microgard™ 2500 Plus PAPR Encapsulated Coveralls

Catalog Number 15536564
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

MICROGARD™ 2500 Plus PAPR Encapsulated Coveralls are designed for use with Powered Air Purifying Respirators (PAPRs) and certified to provide both respiratory and body protection from hazardous substances.

  • Fully encapsulated coverall, for use in combination with Scott Proflow PAPR fan unit, breathing hose and filters
  • Double elasticated cuffs, attached socks with boot over-flap.
  • Emergency rip cord attached to left chest shoulder and left back shoulder as worn
  • Achieves the highest classifications for protection from biological agents in accordance with EN 14126:2003 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
  • Moisture vapour permeable (“breathable”) to help reduce the risk of heat stress
  • Anti-static according to EN 1149-5
  • Low linting for reduced risk of contamination in critical areas
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, Type 6 EN13034, EN14126, EN1073-2, EN1149-1
Color White
Hood Type Fully Encapsulated
Material Microporous Polypropylene Laminate
Size Medium
Ankle Style Attached socks, Boot overflaps
Back Style Accommodates Scott Proflow 2SC/EX PAPR fan unit, breathing hoses, and filters
Cuff Style Double, Elastic
Description Emergency rip cord, attached socks with boot overflap
For Use With (Equipment) Scott Proflow 2SC/EX PAPR fan unit, breathing hoses and filters
No. of Pockets None
Product Type Protective coverall
Seam Type Stitched, Taped
Style Includes Emergency rip cord, attached socks with boot overflap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'