Läs mer
Micronomicin sulfate, 590 ^mg/mg, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar J66072.09
Kvantitet 10g
Produktkod. 15477289
  • 1535.65 SEK / 10g
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • An aminoglycoside antibiotic commonly used against gram negative bacteria
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
Micronomicin sulfate
66803-19-8
10g
C20H43N5O11S
667°C
MQDGQSCLOYLSEK-SCFBDNQUSA-N
(2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(methylamino)methyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; sulfuric acid
171808
590μg/mg
White to Yellow
>110°C (230°F)
MFCD00941451
Freely soluble in water
OS(O)(=O)=O.CNC[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1
561.65
561.65
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer