Läs mer
Ismatec™ Micropump™ GJ Series B-Mount Cavity Gear Pump Head
Flödeshastighet:
0,316 ml/varv
0,91 ml/varv
Förpackningsstorlek:
Varje
Beskrivning
- Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency.
- Cavity style designs with no dynamic seals for applications that cannot tolerate leaks.
- Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery.
- The magnetic drive and static seals keep fluids securely inside and potential contaminants out.
- Chemically resistant materials handle a wide range of materials (due to wetted parts).
- Pump heads are compact and easy to service.
- Modular design offers flexibility—configure to your specific needs.
Specifikationer
Specifikationer
| Beskrivning | Differential pressure: 50 psi |
| Flödeshastighet | 0,316 ml/varv |
| Max. Driftstemperatur (metrisk) | 54°C |
| Material | Gear material: PTFE |
| Material (tätning) | PTFE |
| Min. Driftstemperatur (metrisk) | −46°C |
| Viskositet | 0.5-1500 cP |
| Hastighet | 5500 rpm |
| Material för fodral | Stainless Steel |