Läs mer
DuraLab Microscope Dust Cover for DR & DX Series
Beskrivning
- Maintains a clean, dust-free environment to extend the microscope's lifespan and ensure optimal performance
- Dimensions: H 500 mm x W 400 mm
- Compatible with most microscope brands-verify the size before purchasing
Specifikationer
Specifikationer
| Typ | Dammskydd |
Compatible with DRL0040 - DuraLab DR-410 Advanced Achromatic Monocular Microscope, DRL0041 - DuraLab DR-420 Advanced Achromatic Binocular Microscope, DRL0042 - DuraLab DR-510 Advanced Semi-Plan Monocular Microscope, DRL0043 - DuraLab DR-520 Advanced Semi-Plan Binocular Microscope, DRL0044 - DuraLab DR-530 Advanced Semi-Plan Trinocular Microscope, DRL0045 - DuraLab DX-820 Pro Infinity Binocular Microscope, DRL0046 - DuraLab DX-830 Pro Infinity Trinocular Microscope