missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Wide-field Microscope Eyepiece

Use this eyepiece with compound microscopes. Fisherbrand™ Wide-field Microscope Eyepieces are ergonomically designed for everyday use.

Brand:  Fisherbrand™

Additional Details : Weight : 0.01000kg

Product Code. 12348508

  • 1420.00 SEK / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

For compound microscopes FB69019 and FB69020
  • Affordable compound microscopes for use by students and professionals
  • Includes powerful low voltage illumination with intensity controller, a built in mechanical stage and four
  • achromatic objectives 4x, 10x, 40x and 100x oil (100x for FB69020 only)
  • 10x WF eyepieces
  • Pre-centered Abbe condenser
  • Includes immersion oil 10mL (FB69020 only), blue filter, dust cover and instruction manual
Specifications

Specifications

Widefield Eyepiece
12 mm
Compound microscopes FB69019 and FB69020
15X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers