Learn More
DuraLab Microscope Objective 100x for DF-50
missing translation for 'orderingAttributeHoverText'
Typ:
Microscope Objective
Description
- Provides high magnification for observing cells and microorganisms in detail
- Spring-loaded lens ensures safe handling with immersion oil
- Supports advanced microscopy skills and detailed biological studies
- Aligned with UK science curriculum for in-depth biological exploration and preparation for higher education
- Compatible with 20x eyepiece to further increase total magnification
Specifications
Specifications
| Typ | Microscope Objective |
| För användning med (utrustning) | DRL0038 |
Compatible with DRL0038 - DuraLab DF-50 Classic Monocular Microscope