Learn More
Thermo Scientific™ Microscope Slides, Ground 45°
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Brand:  Thermo Scientific™ ACAC0900130002EA0MNZ#0#G#
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15224788
  • 14290.00 SEK / Pack of 1000
Estimated Shipment
16-12-2020
Add to basket
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Specifications
130mm
1mm
45°, ground
50 pieces in paper
130 x 90mm
90mm
Uncoated
Soda-lime glass