Learn More
Fisherbrand™ Penetration pH Electrodes
Useful for measuring potential of hydrogen ions. Fisherbrand™ Penetration pH Electrodes can be placed deep within samples.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11788462
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Reinforced
  • Excellent for meat, cheese, fish, fruit
  • Electrolyte KCI 3M + AgCl
  • pH range of 0 to 12
  • Temperature range of 0° to 80°C
Specifications
Specifications
DIN
0°C to 80°C
75 mm
0 to 12
Standard
12 mm