Learn More
Labnet™ Microtube Box with Hinged Lid
Ideal for sample storage, polypropylene Microtube Boxes with Hinged Lids are safe for freezer or autoclave.
Brand:  Labnet™ R7901
Additional Details : Weight : 0.01000kg
Product Code. 11426657
  • 72.80 SEK / Pack of 75
Estimated Shipment: 09-12-2022
to see stock.
Add to basket
Description
Description
  • Ideal for sample storage, these polypropylene boxes can be used in the freezer (to -80ºC) or autoclaved.
  • The easy open lid with living hinge and positive catch keeps samples securely in place.
  • The translucent lid is alphanumerically labeled.
  • Boxes available for 1.5/2.0 mL and 0.5 mL tubes.
  • R7901 also holds cryovials and chromotography vials.
Specifications
Specifications
50 x 1.5/2.0 mL
Hinged Lid
Blue
Polypropylene