missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIER2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17426-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Transcriptional repressor.Specifications
MIER2 | |
Polyclonal | |
Unconjugated | |
MIER2 | |
KIAA1193, Mi er2, MIER2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362841, 54531, 70427 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3U3N0, Q8N344 | |
Mier2 | |
MIER2 Fusion Protein Ag11491 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |