Läs mer
Miglustat hydrochloride, MedChemExpress
Produktkod. 16479787
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
Förpackningsstorlek:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16479787 5 mg -
16487268 10 mg -
16453566 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16479787 Leverantör Medchem Express Leverantörsnummer HY17020A
MedChemExpress Miglustat hydrochloride is an inhibitor of glucosylceramide synthase, primarily to treat Type I Gaucher disease (GD1).
Miglustat hydrochloride is an inhibitor of glucosylceramide synthase, primarily to treat Type I Gaucher disease (GD1).
Specifikationer
Kemiskt namn eller material Miglustat hydrochloride
Fysisk form Oil
CAS 210110-90-0
Färg Light Brown
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C10H22ClNO4
För användning med (applikation) Neuroscience-Neuromodulation
Synonym N-Butyldeoxynojirimycin hydrochlorideNB-DNJ hydrochlorideOGT 918 hydrochloride
Löslighetsinformation DMSO : 65 mg/mL (254.16 mM; Need ultrasonic) ∣H2O : ≥ 34 mg/mL (132.95 mM)
LEDER O[C@H]1[C@H](O)[C@@H](CO)N(CCCC)C[C@@H]1O.[H]Cl
Molekylvikt (g/mol) 255.74
Formel vikt 255.74
Procent renhet 98.0%