Läs mer
Milbemycin D (~90%), TRC
Produktkod. 30478771
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30478771 1 mg
1 options
Produktkod. 30478771 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3443051MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Milbemycin D
CAS 77855-81-3
Molekylformel C33H48O7
Kvantitet 1 mg
Synonym Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran], Antibiotic B 41D, Milbemycin EP Impurity C
InChI formel InChI=1S/C33H48O7/c1-19(2)29-22(5)12-13-32(40-29)17-26-16-25(39-32)11-10-21(4)14-20(3)8-7-9-24-18-37-30-28(34)23(6)15-27(31(35)38-26)33(24,30)36/h7-10,15,19-20,22,25-30,34,36H,11-14,16-18H2,1-6H3/b8-7+,21-10+,24-9+/t20-,22-,25+,26-,27-,28+,29+,30+,32+,33+/m0/s1
LEDER CC(C)[C@H]1O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(/C)\C[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O3)[C@]45O)C)O2
Molekylvikt (g/mol) 556.73
Formel vikt 556.34