Läs mer
MilliporeSigma Millicell™ ERS 3.0 Digital Voltohmmeter
Produktkod. 18844620 Handla allt Merck Produkter
Change view
Spänning:
0/1 V
Produktkod. Spänning
18844620 0/1 V
1 options
Produktkod. 18844620 Leverantör MilliporeSigma Leverantörsnummer MERS03000
  • ONLINE EXCLUSIVE PRICE
    72555.00 SEK
    66169.01 SEK / Paket med 1
    Save 6385.99 SEK
    8% Av
Visa alla kampanjer
 Begränsad artikel
Beräknad leverans: 27-11-2026
The Millicell Electrical Resistance System reliably measures transmembrane potential and resistance of endothelial/epithelial cells in culture.
  • Reliably measures membrane potential and resistance of epithelial and endothelial cells in culture
  • Qualitatively measures cell monolayer health and cell confluence
  • An Electroless Nickle Immersion Gold (ENIG) plate on each external facing electrode tip measures voltage
  • Small electrode size enables easy measurement of transepithelial voltage and the resistance of cells grown on microporous membranes
  • Comes with the Voltohmmeter, standard adjustable electrode, a Wi-Fi USB dongle, a power cord, a verification device and free 6-month trial subscription to Millicell™ Cloud account.
  • Each instrument is battery compatible but can run off only AC wall power in the absence of a battery.
TRUSTED_SUSTAINABILITY
Specifikationer
Typ Digital Voltohmmeter
Spänning 0/1 V