Learn More
Merck Accessories for 47 mm All-glass Filter Holders Kits and MilliSolve Kit
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Brand: Merck XX1514732
4920.00 SEK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.54680kg
Description
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Specifications
Clear | |
Stainless Steel | |
Stainless steel screen | |
47 mm |
Millipore all-glass filter holders kits | |
Vacuum filtration | |
Glass base and tubulated cap |