Läs mer
Milrinone, TRC
Produktkod. 30134161
Change view
Kvantitet:
5 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134161 250 mg
30136629 100 mg
18081620 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134161 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM344680250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 78415-72-2
Molekylformel C12 H9 N3 O
Molekylvikt (g/mol) 211.22
Synonym Milrinone, [3,4'-Bipyridine]-5-carbonitrile, 1,6-dihydro-2-methyl-6-oxo-, 1,6-Dihydro-2-methyl-6-oxo-(3,4'-bipyridine)-5-carbonitrile, Corotrop, Corotrope, Milrinone, Primacor IV, Win 47203
IUPAC-namn 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carbonitrile
LEDER CC1=C(C=C(C#N)C(=O)N1)c2ccncc2
Specifikationer
Kemiskt namn eller material Milrinone
Kvantitet 250 mg
InChI formel InChI=1S/C12H9N3O/c1-8-11(9-2-4-14-5-3-9)6-10(7-13)12(16)15-8/h2-6H,1H3,(H,15,16)
Formel vikt 211.0746