Läs mer
Miltefosine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Miltefosine |
| CAS | 58066-85-6 |
| Molekylformel | C21 H46 N O4 P |
| Kvantitet | 50 mg |
| Synonym | Ethanaminium, 2-[[(hexadecyloxy)hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt, Choline phosphate, hexadecyl ester, hydroxide, inner salt (6CI), D 18506, Fos-choline 16, HPC, Hexadecylphosphocholine, Hexadecylphosphorylcholine, Impavido, Miltefos, Miltefosine, Miltex, NSC 605583, n-Hexadecylphosphocholine, n-Hexadecylphosphorylcholine |
| InChI formel | InChI=1S/C21H46NO4P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-20-25-27(23,24)26-21-19-22(2,3)4/h5-21H2,1-4H3 |
| LEDER | CCCCCCCCCCCCCCCCOP(=O)([O-])OCC[N+](C)(C)C |
| IUPAC-namn | hexadecyl 2-(trimethylazaniumyl)ethyl phosphate |
| Molekylvikt (g/mol) | 407.57 |
| Formel vikt | 407.3164 |