Learn More
Ohaus™ Mini Fix Speed Vortex Mixer VXMNFS
OHAUS™ Mini Fix Speed Vortex Mixer VXMNFS is built sturdy to provide a stable and reliable vortexing action.
Brand:  Ohaus™ 30392110
Additional Details : Weight : 4.19400kg
Product Code. 15782229
  • 2960.00 SEK / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • Sturdy design and construction to provide stable and reliable vortexing action
  • Starts mixing when the cup head is depressed with touch-mode operation
  • Speed is fixed at full rpm to provide vigorous vortexing of samples
Specifications
Specifications
4.9 mm
230 V, 50/60 Hz, 0.6 A
Mini Fix Speed Vortex Mixer
21.00 x 12.30 cm
16.5 cm
21.00 cm
12.30 cm
2500 rpm
0.6 A
EU Plug
6.50 in.
8.30 in.
4.80 in.
VXMNFS, EU
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote