Learn More
Trajan Scientific and Medical™ Mini Union Replacement Ferrules
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
Brand: Trajan Scientific and Medical™ 072694
Additional Details : Weight : 0.01000kg
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
Specifications
Bore size: No Hole | |
15% Graphite, 85% Vespel | |
Graphite Vespel Replacement Mini Union Ferrule No-Hole | |
10/Pk. |