missing translation for 'onlineSavingsMsg'
Learn More

MIPOL1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14158-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16822674

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a coiled-coil domain-containing protein. The encoded protein may function as a tumor suppressor. A translocation that results in truncation of the protein encoded by this locus has been associated with mirror-image polydactyly, also known as Laurin-Sandrow Syndrome. Alternatively spliced transcript variants have been described.
Specifications

Specifications

MIPOL1
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8TD10
MIPOL1, mirror image polydactyly 1
Rabbit
IgG
150 μL
-20°C
Primary
145282
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MIPOL1
Liquid
MIPOL1
MIPOL1 Fusion Protein Ag5340
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers