Läs mer
Tocris Bioscience™ MIRA-1
GREENER_CHOICE
Produktkod. 18004532
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18004532 10 mg
18044962 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18004532 Leverantör Tocris Bioscience™ Leverantörsnummer 3362/10
Restores mutant p53 activity; proapoptotic
Restores wild-type conformation, function and DNA binding activity to mutant p53. Induces p53 transcriptional transactivation of p21, MDM2 and PUMA, and promotes tumor cell death by apoptosis in a mutant p53-dependent manner in vitro (IC50 = 10μM).
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72835-26-8
Molekylformel C8H9NO4
Specifikationer
Kemiskt namn eller material MIRA-1
Kvantitet 10 mg
Formel vikt 183.16
Renhet >98%
Molekylvikt (g/mol) 183.163