Läs mer
Miramistin, TRC
Produktkod. 17907704
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17907704 250 mg
17940075 500 mg
17939334 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17907704 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM365035250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15809-19-5
Molekylformel C26 H47 N2 O . Cl
Molekylvikt (g/mol) 439.12
Synonym N,N-Dimethyl-N-[3-[(1-oxotetradecyl)amino]propyl]-benzenemethanaminium Chloride, Benzyldimethyl(3-tetradecamidopropyl)ammonium Chloride, Benzyldimethyl[3-(myristoylamino)propyl]ammonium Chloride, Myramistin, Benzyldimethyl(3-myristamidopropyl)-ammonium Chloride, DT 2, DT 2 (surfactant);
IUPAC-namn benzyl-dimethyl-[3-(tetradecanoylamino)propyl]ammonium;chloride
LEDER [Cl-].CCCCCCCCCCCCCC(=O)NCCC[N+](C)(C)Cc1ccccc1
Specifikationer
Kemiskt namn eller material Miramistin
Kvantitet 250 mg
InChI formel InChI=1S/C26H46N2O.ClH/c1-4-5-6-7-8-9-10-11-12-13-17-21-26(29)27-22-18-23-28(2,3)24-25-19-15-14-16-20-25;/h14-16,19-20H,4-13,17-18,21-24H2,1-3H3;1H
Formel vikt 438.3377