Läs mer
Mirtazapine Bromide, TRC
Produktkod. 30480610
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30480610 5 mg
1 options
Produktkod. 30480610 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3650055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mirtazapine Bromide
CAS 61337-86-8
Molekylformel C17H18BrN3
Kvantitet 5 mg
Synonym 8-Bromo Mirtazapine, 8-Bromo-1,2,3,4,10,14b-hexahydro-2-methyl-pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine.
InChI formel InChI=1S/C17H18BrN3/c1-20-6-7-21-16(11-20)15-5-3-2-4-12(15)8-13-9-14(18)10-19-17(13)21/h2-5,9-10,16H,6-8,11H2,1H3
LEDER CN1CCN2C(C1)c3ccccc3Cc4cc(Br)cnc24
Molekylvikt (g/mol) 344.25
Formel vikt 343.0684