Learn More
R&D Systems™ Mito-HE
Product Code. 18729466
Change view
Click to view available options
Quantity:
1 Pk. (10 x 50 μg)
Unit Size:
1 set
Product Code. Quantity Unit Size
18729466 1 Pk. (10 x 50 μg) 1 set
1 options
Product Code. 18729466 Supplier R&D Systems™ Supplier No. 7641/1 PACK
  • ONLINE EXCLUSIVE PRICE
    4182.71 SEK
    3726.45 SEK / 1 uppsättningar
    Save 456.26 SEK
    10% Av
 Restricted Item
Beräknad leverans: 27-11-2026
A Tocris product, part of R&D Systems. Fluorescent mitochondrial superoxide indicator in live cells
Mito-HE is a fluorescent probe for imaging superoxide in mitochondria of living cells. Selective for mitochondrial superoxides over other reactive oxygen and nitrogen species. Can be used to detect mitochondrial superoxide in Parkinson's and Alzheimer's disease in vitro models. Excitation/emission maxima λ ∼ 510/580 nm.
It is recommended to prepare stock solutions in DMSO.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material [6-(3,8-Diamino-6-phenyl-5(6 H)-phenanthridinyl)hexyl]triphenylphosphonium iodide
CAS 1003197-00-9
Quantity 1 Pk. (10 x 50 μg)
Product Line Tocris
Target Mitochondria Dyes and Probes
Molecular Formula C43H43IN3P
Purity 0.8