missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitoferrin 1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26469-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
SLC25A37 belongs to the SLC25 family of mitochondrial carrier proteins.Specifications
Mitoferrin 1 | |
Polyclonal | |
Unconjugated | |
SLC25A37 | |
MFRN, Mfrn1, Mitochondrial iron transporter 1, Mitoferrin 1, SLC25A37 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51312, 67712 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q920G8, Q9NYZ2 | |
SLC25A37 | |
Mitoferrin 1 Fusion Protein Ag24876 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |