Läs mer
Mitoquinol, TRC
Produktkod. 30483216
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483216 500 mg
30469908 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30483216 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM372210500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 845959-55-9
Molekylformel C38H49O7PS
Molekylvikt (g/mol) 680.83
Synonym [10-(3,6-Dihydroxy-4,5-dimethoxy-2-methylphenyl)decyl]triphenylphosphonium Methanesulfonate, 10-(2,5-Dihydroxy-3,4-dimethoxy-6-methylphenyl)decyl]triphenylphosphonium Methanesulfonate (1:1)
LEDER OC1=C(OC)C(OC)=C(O)C(C)=C1CCCCCCCCCC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.CS(=O)([O-])=O
Specifikationer
Kemiskt namn eller material Mitoquinol
Kvantitet 500 mg
InChI formel InChI=1S/C37H45O4P.CH4O3S/c1-29-33(35(39)37(41-3)36(40-2)34(29)38)27-19-8-6-4-5-7-9-20-28-42(30-21-13-10-14-22-30,31-23-15-11-16-24-31)32-25-17-12-18-26-32;1-5(2,3)4/h10-18,21-26H,4-9,19-20,27-28H2,1-3H3,(H-,38,39);1H3,(H,2,3,4)