Läs mer
Mitoxantrone, TRC
Produktkod. 30473139
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30473139 500 mg
1 options
Produktkod. 30473139 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM373425500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mitoxantrone
CAS 65271-80-9
Molekylformel C22 H28 N4 O6
Kvantitet 500 mg
Synonym 9,10-Anthracenedione, 1,4-dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]-, 1,4-Dihydroxy-5,8-bis[[2-[(2-hydroxyethyl)amino]ethyl]amino]-9,10-anthracenedione, 1,4-Bis[(2-(2-hydroxyethylamino)ethyl)amino]-5,8-dihydroxyanthraquinone, 1,4-Dihydroxy-5,8-bis-[[2-[(2-hydroxyethyl)amino]ethyl]amino]anthraquinone, DHAQ, Dihydroxyanthraquinone, Mitox, Mitoxanthrone, Mitoxantrone, Mitozantrone, NSC 279836, Nimitoxantron, Novantron, Novantrone, Ralenova
InChI formel InChI=1S/C22H28N4O6/c27-11-9-23-5-7-25-13-1-2-14(26-8-6-24-10-12-28)18-17(13)21(31)19-15(29)3-4-16(30)20(19)22(18)32/h1-4,23-30H,5-12H2
LEDER OCCNCCNc1ccc(NCCNCCO)c2C(=O)c3c(O)ccc(O)c3C(=O)c12
IUPAC-namn 1,4-dihydroxy-5,8-bis[2-(2-hydroxyethylamino)ethylamino]anthracene-9,10-dione
Molekylvikt (g/mol) 444.48
Formel vikt 444.2009