Läs mer
Mitragynine, 1mg/ml in Methanol, Lipomed
Produktkod. 30463593
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30463593 1 ml
1 options
Produktkod. 30463593 Leverantör Lipomed Leverantörsnummer LPMMTR16101LM
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Mitragynine, 1mg/ml in Methanol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mitragynine
CAS 4098-40-2
Analyt- eller komponentnamn Mitragynine
Frakt skick Room Temperature
Molekylformel C23 H30 N2 O4
Koncentration 1 mg/mL
Synonym Mitragynine
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER CC[C@@H]1 CN2CCc3c([nH]c4cccc(OC)c34)[C@@H]2 C[C@@H]1 \C(=C/OC)\C(=O)OC
IUPAC-namn methyl (E)-2-[(2 S,3 S,12 bS)-3-ethyl-8-methoxy-1,2,3,4,6,7,12,12 b-octahydroindolo[2,3-a]quinolizin-2-yl]-3-methoxyprop-2-enoate
Lösningsmedel eller matris Methanol
Industry Type Pharmaceutical
Molekylvikt (g/mol) 398.5
Kvantitet 1 ml
Formel vikt 398.2206 g/mol