Läs mer
Mitragynine, Lipomed
Produktkod. 30458788
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30458788 50 mg
1 options
Produktkod. 30458788 Leverantör Lipomed Leverantörsnummer LPMMTR161050
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Mitragynine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mitragynine
CAS 4098-40-2
Analyt- eller komponentnamn Mitragynine
Frakt skick Room Temperature
Molekylformel C23 H30 N2 O4
Synonym Mitragynine
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER CC[C@@H]1 CN2CCc3c([nH]c4cccc(OC)c34)[C@@H]2 C[C@@H]1 \C(=C/OC)\C(=O)OC
IUPAC-namn methyl (E)-2-[(2 S,3 S,12 bS)-3-ethyl-8-methoxy-1,2,3,4,6,7,12,12 b-octahydroindolo[2,3-a]quinolizin-2-yl]-3-methoxyprop-2-enoate
Industry Type Pharmaceutical
Molekylvikt (g/mol) 398.5
Kvantitet 50 mg
Formel vikt 398.2206 g/mol