Läs mer
Mitragynine, Lipomed
Produktkod. 30460917
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30460917 100 mg
1 options
Produktkod. 30460917 Leverantör Lipomed Leverantörsnummer LPMMTR1610100
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Mitragynine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mitragynine
CAS 4098-40-2
Analyt- eller komponentnamn Mitragynine
Frakt skick Room Temperature
Molekylformel C23 H30 N2 O4
Synonym Mitragynine
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER CC[C@@H]1 CN2CCc3c([nH]c4cccc(OC)c34)[C@@H]2 C[C@@H]1 \C(=C/OC)\C(=O)OC
IUPAC-namn methyl (E)-2-[(2 S,3 S,12 bS)-3-ethyl-8-methoxy-1,2,3,4,6,7,12,12 b-octahydroindolo[2,3-a]quinolizin-2-yl]-3-methoxyprop-2-enoate
Industry Type Pharmaceutical
Molekylvikt (g/mol) 398.5
Kvantitet 100 mg
Formel vikt 398.2206 g/mol