Läs mer
Mitragynine, TRC
Produktkod. 30477376
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30477376 5 mg
1 options
Produktkod. 30477376 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3735505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mitragynine
CAS 4098-40-2
Molekylformel C23 H30 N2 O4
Kvantitet 5 mg
Synonym Indolo[2,3-a]quinolizine-2-acetic acid, 3-ethyl-1,2,3,4,6,7,12,12b-octahydro-8-methoxy-α-(methoxymethylene)-, methyl ester, (αE,2S,3S,12bS)-, Corynan-16-carboxylic acid, 16,17-didehydro-9,17-dimethoxy-, methyl ester, (16E,20β)-, Corynantheidine, 9-methoxy- (7CI), Mitragynine (6CI), (-)-Mitragynine, 9-Methoxycorynantheidine, Indolo[2,3-a]quinolizine-2-acetic acid, 3-ethyl-1,2,3,4,6,7,12,12b-octahydro-8-methoxy-α-(methoxymethylene)-, methyl ester, [2S-[2α(E),3α,12bβ]]-, Mitragynin, Kratom
InChI formel InChI=1S/C23H30N2O4/c1-5-14-12-25-10-9-15-21-18(7-6-8-20(21)28-3)24-22(15)19(25)11-16(14)17(13-27-2)23(26)29-4/h6-8,13-14,16,19,24H,5,9-12H2,1-4H3/b17-13+/t14-,16+,19+/m1/s1
LEDER CC[C@@H]1CN2CCc3c([nH]c4cccc(OC)c34)[C@@H]2C[C@@H]1\C(=C/OC)\C(=O)OC
IUPAC-namn methyl (E)-2-[(2S,3S,12bS)-3-ethyl-8-methoxy-1,2,3,4,6,7,12,12b-octahydroindolo[2,3-a]quinolizin-2-yl]-3-methoxyprop-2-enoate
Molekylvikt (g/mol) 398.5
Formel vikt 398.2206