Learn More
Mixed Calibration Standard 3 For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
1870.00 SEK - 2240.00 SEK
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11941646
|
SPEX Certiprep
MIXSTD3-100 |
125 mL | Triple-leached LDPE bottle |
1870.00 SEK
125mL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11951646
|
SPEX Certiprep
MIXSTD3-500 |
500 mL | Triple-leached LDPE bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As | 500μg/mL |
Mo | 100μg/mL |
Si | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Product intended for laboratory use only