missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIXL1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22772-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to MIXL1.
MIXL1 is a novel human Mix-like homeobox gene. In normal hematopoiesis, its expression appears to be restricted to immature B- and T-lymphoid cells.Specifications
MIXL1 | |
Polyclonal | |
Unconjugated | |
MIXL1 | |
hMix, Homeobox protein MIXL1, Homeodomain protein MIX, MILD1, MIX, Mix.1 homeobox like protein, MIX1 homeobox like protein 1, MIXL, MIXL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
289311, 83881 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H2W2 | |
Mixl1 | |
MIXL1 Fusion Protein Ag18732 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |