Läs mer
Mixture of (-)-Angustifoline and Isoangustifoline, TRC
Produktkod. 30012628
Change view
Kvantitet:
0.5 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012628 0.5 mg
30003214 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012628 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6378900.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 550-43-6
Molekylformel C14 H22 N2 O
Molekylvikt (g/mol) 234.3373
Synonym 1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, decahydro-4-(2-propen-1-yl)-, (1S,4S,5S,11aR)-, 1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, decahydro-4-(2-propenyl)-, (1S,4S,5S,11aR)- (9CI), 1,5-Methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, decahydro-4-(2-propenyl)-, [1S-(1α,4α,5α,11aα)]-, Angustifoline (6CI,7CI,8CI), (1S,4S,5S,11aR)-Decahydro-4-(2-propen-1-yl)-1,5-methano-8H-pyrido[1,2-a][1,5]diazocin-8-one, (-)-Angustifoline, Jamaicensine
IUPAC-namn (1S,2R,9S,10S)-10-prop-2-enyl-7,11-diazatricyclo[7.3.1.02,7]tridecan-6-one
LEDER C=CC[C@@H]1NC[C@@H]2C[C@H]1CN3[C@@H]2CCCC3=O
Specifikationer
Kemiskt namn eller material (-)-Angustifoline
Analyt- eller komponentnamn (-)-Angustifoline
Frakt skick Room Temperature
InChI formel InChI=1S/C14H22N2O/c1-2-4-12-11-7-10(8-15-12)13-5-3-6-14(17)16(13)9-11/h2,10-13,15H,1,3-9H2/t10-,11-,12-,13+/m0/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 0.5 mg
Formel vikt 234.1732