Läs mer
Tocris Bioscience™ MK 0677
High affinity ghrelin receptor agonist
Brand: Tocris Bioscience™ 5272/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MK 0677 is a high affinity ghrelin receptor agonist (pKi = 8.14). Growth hormone (GH) secretagog; stimulates GH release from rat pituitary cells in vitro (EC50 = 1.3 nM) and enhances GH plasma levels in vivo. Also attenuates isoproterenol -induced lipolysis in rat adipocytes in vitro. Orally bioavailable.Specifikationer
| 2-Amino-N-[(1 R)-2-[1,2-dihydro-1-(methylsulfonyl)spiro[3 H-indole-3,4 '-piperidin]-1 '-yl]-2-oxo-1-[(phenylmethoxy)methyl]ethyl]-2-methylpropanamide methanesulfonate | |
| 159752-10-0 | |
| Ibutamoren mesylate, L-163,191 | |
| 10 mg |