Läs mer
MK-0822, TRC
Produktkod. 18062620
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18062620 100 mg
18050020 50 mg
18072820 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18062620 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM425015100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 603139-19-1
Molekylformel C25 H27 F4 N3 O3 S
Molekylvikt (g/mol) 525.5588
Synonym MK-0822;N-(1-cyanocyclopropyl)-4-fluoro-N2-{(1S)-2,2,2-trifluoro-1-[4'-(methylsulfonyl)biphenyl-4-yl]ethyl}-L-leucinamide, Odanacatib
IUPAC-namn (2S)-N-(1-cyanocyclopropyl)-4-fluoro-4-methyl-2-[[(1S)-2,2,2-trifluoro-1-[4-(4-methylsulfonylphenyl)phenyl]ethyl]amino]pentanamide
LEDER CC(C)(F)C[C@H](N[C@@H](c1ccc(cc1)c2ccc(cc2)S(=O)(=O)C)C(F)(F)F)C(=O)NC3(CC3)C#N
Specifikationer
Kemiskt namn eller material (2S)-N-(1-cyanocyclopropyl)-4-fluoro-4-methyl-2-[((1S)-2,2,2-trifluoro-1-[4-(4-methylsulfonylphenyl)phenyl]ethyl)amino]pentanamide
InChI formel InChI=1S/C25H27F4N3O3S/c1-23(2,26)14-20(22(33)32-24(15-30)12-13-24)31-21(25(27,28)29)18-6-4-16(5-7-18)17-8-10-19(11-9-17)36(3,34)35/h4-11,20-21,31H,12-14H2,1-3H3,(H,32,33)/t20-,21-/m0/s1
Kvantitet 100 mg
Formel vikt 525.1709