Läs mer
MK-3207 Hydrochloride, MedChemExpress
Produktkod. 16496497
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16496497 5 mg 5mg
16460157 50 mg -
16453778 10 mg -
16434836 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496497 Leverantör Medchem Express Leverantörsnummer HY10302
MedChemExpress MK-3207 (Hydrochloride) is a potent and orally bioavailable CGRP receptor antagonist with IC50 of 0.12 nM and Ki of 0.024 nM, and is highly selective versus human AM1, AM2, CTR, and AMY3.
MK-3207 (Hydrochloride) is a potent and orally bioavailable CGRP receptor antagonist with IC50 of 0.12 nM and Ki of 0.024 nM, and is highly selective versus human AM1, AM2, CTR, and AMY3.
Specifikationer
Kemiskt namn eller material MK-3207 Hydrochloride
CAS 957116-20-0
Färg Pink
Anteckningar om renhetsgrad Research
Molekylformel C31H30ClF2N5O3
Kvantitet 5 mg
Löslighetsinformation DMSO : ≥ 100 mg/mL (168.34 mM) ∣H2O : 50 mg/mL (84.17 mM; Need ultrasonic)
LEDER O=C1C2(NC[C@@H](C3=CC(F)=CC(F)=C3)N1CC(NC4=CC5=C(C[C@@]6(C(NC7=C6C=CC=N7)=O)C5)C=C4)=O)CCCC2.Cl
Molekylvikt (g/mol) 594.05
Formel vikt 594.05
Procent renhet 99.06%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation