Läs mer
MK-571, TRC
Produktkod. 30465850
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30465850 5 mg
30474594 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30465850 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM4245005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115104-28-4
Molekylformel C26 H26 Cl N2 O3 S2 . Na
Molekylvikt (g/mol) 537.07
Synonym MK 571
IUPAC-namn sodium;3-[[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-[3-(dimethylamino)-3-oxopropyl]sulfanylmethyl]sulfanylpropanoate
LEDER [Na+].CN(C)C(=O)CCSC(SCCC(=O)[O-])c1cccc(\C=C\c2ccc3ccc(Cl)cc3n2)c1
Specifikationer
Kemiskt namn eller material MK-571
Kvantitet 5 mg
InChI formel InChI=1S/C26H27ClN2O3S2.Na/c1-29(2)24(30)12-14-33-26(34-15-13-25(31)32)20-5-3-4-18(16-20)6-10-22-11-8-19-7-9-21(27)17-23(19)28-22;/h3-11,16-17,26H,12-15H2,1-2H3,(H,31,32);/q;+1/p-1/b10-6+;
Formel vikt 536.0971